accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15366-34-4

CH3OONHN

CAS RN 15366-34-4

IUPAC Name: methyl 1H-pyrazole-5-carboxylate
Synonyms: methyl 2H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: ORUCTBNNYKZMSK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results