CAS RN 153766-81-5
IUPAC Name:
potassium trifluoro(phenyl)boranuide
Synonyms:
potassium trifluoro(phenyl)boranuide
Molecular Weight (g/mol):
184.01
Molecular Formula:
C6H5BF3K
InChi Key:
DVAFPKUGAUFBTJ-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC=CC=C1
1
–
1
of
1
results
1
Potassium phenyltrifluoroborate, 98%
CAS: 153766-81-5 Molecular Formula: C6H5BF3K Molecular Weight (g/mol): 184.01 MDL Number: MFCD01318172 InChI Key: DVAFPKUGAUFBTJ-UHFFFAOYSA-N Synonym: potassium phenyltrifluoroborate,potassium trifluoro phenyl borate,potassium trifluoro phenyl boranuide,pubchem11425,potassiumphenyltrifluoroborate,amtb100,phenyltrifluoropotassioboron v,potassium trifluoro phenyl borate 1-,potassium ion trifluoro phenyl boranuide,potassium tris fluoranyl-phenyl-boranuide PubChem CID: 23675248 IUPAC Name: potassium;trifluoro(phenyl)boranuide SMILES: [B-](C1=CC=CC=C1)(F)(F)F.[K+]