accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOFFNOO

CAS RN 153775-33-8

IUPAC Name: 2,4-difluoro-5-nitrobenzoate
Synonyms: 2,4-difluoro-5-nitrobenzoate
Molecular Weight (g/mol): 202.09
Molecular Formula: C7H2F2NO4
InChi Key: MDFSGDMPHMKKGB-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results