accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHNNOOOHO

CAS RN 1539-06-6

IUPAC Name: 4-acetamido-3-nitrobenzoic acid
Synonyms: 4-acetamido-3-nitrobenzoic acid
Molecular Weight (g/mol): 224.17
Molecular Formula: C9H8N2O5
InChi Key: BRQIMWBIZLRLSV-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results