accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 153912-60-8

IUPAC Name: (1,5-dimethyl-1H-pyrazol-3-yl)methanol
Synonyms: (1,5-dimethylpyrazol-3-yl)methanol
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: XJBMHIHXRARJJS-UHFFFAOYSA-N
SMILES: CN1N=C(CO)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results