CAS RN 154-02-9
CAS RN 154-02-9
IUPAC Name:
3-(2-hydroxyethyl)-1H-indol-5-ol
Synonyms:
hydroxytryptophol 5-htol 5-hydroxy-3-indole 5-hydroxytryptophol 5-hydroxyindolacetate acid 5-hydroxyindoacetic acid
Molecular Weight (g/mol):
177.2
Molecular Formula:
C10H11NO2
InChi Key:
KQROHCSYOGBQGJ-UHFFFAOYSA-N
SMILES:
OCCC1=CNC2=CC=C(O)C=C12
1
–
1
of
1
results
1
5-Hydroxy Tryptophol, TRC
CAS: 154-02-9 Molecular Formula: C10 H11 N O2 Molecular Weight (g/mol): 177.2 Synonym: 1H-Indole-3-ethanol, 5-hydroxy-,Indole-3-ethanol, 5-hydroxy- (7CI,8CI),Tryptophol, 5-hydroxy- (6CI),5-Hydroxy-1H-indole-3-ethanol,3-(2-Hydroxyethyl)-1H-indol-5-ol,5-Hydroxy-3-(2-hydroxyethyl)indole,5-Hydroxyindole-3-(2-hydroxy)ethyl,5-Hydroxytryptophol,5-Hydroxy tryptophol,NSC 84416,2-(5-Hydroxyindol-3-yl)ethanol IUPAC Name: 3-(2-hydroxyethyl)-1H-indol-5-ol SMILES: OCCc1c[nH]c2ccc(O)cc12