accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 154-58-5

IUPAC Name: (2R,3S,4R,5S)-2-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: 1,5-anhydroglucitol anhydroglucitol
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: MPCAJMNYNOGXPB-SLPGGIOYSA-N
SMILES: OC[C@H]1OC[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results