accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 154-93-8

ClNHONClNO

CAS RN 154-93-8

IUPAC Name: 1,3-bis(2-chloroethyl)-3-nitrosourea
Synonyms: carmustine bcnu (bis chloroethylnitrosourea) carmustinum FIVB bis(chloroethyl) nitrosourea bcnu - carmustine C5H9cl2N3O2 carmustine preparation camustine bicnu (TN)
Molecular Weight (g/mol): 214.05
Molecular Formula: C5H9Cl2N3O2
InChi Key: DLGOEMSEDOSKAD-UHFFFAOYSA-N
SMILES: ClCCNC(=O)N(CCCl)N=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results