accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NONH2

CAS RN 154016-48-5

IUPAC Name: 1-(5-methyl-1,2-oxazol-3-yl)methanamine
Synonyms: 1-(5-methyl-1,2-oxazol-3-yl)methanamine
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: AZVWIMLQRLKLHH-UHFFFAOYSA-N
SMILES: CC1=CC(CN)=NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results