accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

LiCH3NCH3AlNH3CH3CNH3CH3CNCH3CH3

CAS RN 15405-86-4

IUPAC Name: lithium(1+) tetrakis(dibutylamino)alumanuide
Synonyms: lithium(1+) tetrakis(dibutylamino)alumanuide
Molecular Weight (g/mol): 546.88
Molecular Formula: C32H72AlLiN4
InChi Key: ISBVOXPIPNIBOM-UHFFFAOYSA-N
SMILES: [Li+].CCCCN(CCCC)[Al-](N(CCCC)CCCC)(N(CCCC)CCCC)N(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results