accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 154149-38-9

H3CNCH3NNHClHCl(E/Z)

CAS RN 154149-38-9

IUPAC Name: N,N-dimethyl-4-({3,4,5,6-tetrahydro-[2,3'-bipyridin]-3-ylidene}methyl)aniline dihydrochloride
Synonyms: 4-{5,6-dihydro-4H-[2,3'-bipyridin]-3-ylidenemethyl}-N,N-dimethylaniline dihydrochloride
Molecular Weight (g/mol): 364.31
Molecular Formula: C19H23Cl2N3
InChi Key: DHCNEWWWKHBASC-UHFFFAOYSA-N
SMILES: Cl.Cl.CN(C)C1=CC=C(C=C2CCCN=C2C2=CN=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results