accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOBr

CAS RN 154264-95-6

IUPAC Name: 7-bromo-4-methyl-3,4-dihydro-2H-1,4-benzoxazine
Synonyms: 7-bromo-4-methyl-2,3-dihydro-1,4-benzoxazine
Molecular Weight (g/mol): 228.09
Molecular Formula: C9H10BrNO
InChi Key: MQMFOFZKZBLSAB-UHFFFAOYSA-N
SMILES: CN1CCOC2=C1C=CC(Br)=C2
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)