accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2HC

CAS RN 15430-52-1

IUPAC Name: hydrogen prop-2-yn-1-amine chloride
Synonyms: hydrogen 2-propynylamine chloride
Molecular Weight (g/mol): 91.54
Molecular Formula: C3H6ClN
InChi Key: IKXNIQJDNKPPCH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC#C
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)