CAS RN 154356-96-4
CAS RN 154356-96-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(4-Chlorophenyl)[4-(1-methylethoxy)phenyl]methanone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
(4-Chlorophenyl)[4-(1-methylethoxy)phenyl]methanone(Fenofibrate Impurity), TRC
CAS: 154356-96-4 Molecular Formula: C16 H15 Cl O2 Molecular Weight (g/mol): 274.74 Synonym: (4-Chlorophenyl)[4-(1-methylethoxy)phenyl]methanone,Fenofibrate Imp. F (EP) IUPAC Name: (4-chlorophenyl)-(4-propan-2-yloxyphenyl)methanone SMILES: CC(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2