accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15442-57-6

IUPAC Name: platinum(2+) bis((ethylsulfanyl)ethane) dichloride
Synonyms: platinum(2+) bis(ethyl sulfide) dichloride
Molecular Weight (g/mol): 446.35
Molecular Formula: C8H20Cl2PtS2
InChi Key: DCIVLRIRJUWCIS-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pt++].CCSCC.CCSCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results