CAS RN 154801-74-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ent-Efavirenz, TRC
CAS: 154801-74-8 Molecular Formula: C14 H9 Cl F3 N O2 Molecular Weight (g/mol): 315.68 IUPAC Name: (4R)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one SMILES: FC(F)(F)[C@@]1(OC(=O)Nc2ccc(Cl)cc12)C#CC3CC3