accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOOH3CCH3H3CH3C(E/Z)

CAS RN 154820-97-0

IUPAC Name: 4,4,5,5-tetramethyl-2-(5-phenylpent-1-en-1-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(5-phenylpent-1-en-1-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 272.2
Molecular Formula: C17H25BO2
InChi Key: HLUHQMVNJDKHRO-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C=CCCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results