accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOHNOO

CAS RN 15485-80-0

IUPAC Name: 7-hydroxy-3-(4-nitrophenyl)-4H-chromen-4-one
Synonyms: 7-hydroxy-3-(4-nitrophenyl)chromen-4-one
Molecular Weight (g/mol): 283.24
Molecular Formula: C15H9NO5
InChi Key: CEESSYSSRMOMDE-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(=O)C(=COC2=C1)C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results