accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONNNHOCH3H3CHNOCH3H3CNHOO

CAS RN 155026-49-6

IUPAC Name: benzyl N-[1-({1-[(1-diazo-2-oxoheptan-3-yl)carbamoyl]-2-methylpropyl}carbamoyl)-2-methylpropyl]carbamate
Synonyms: benzyl N-[1-({1-[(1-diazo-2-oxoheptan-3-yl)carbamoyl]-2-methylpropyl}carbamoyl)-2-methylpropyl]carbamate
Molecular Weight (g/mol): 487.6
Molecular Formula: C25H37N5O5
InChi Key: IIPZPLSBQVJCBO-UHFFFAOYNA-N
SMILES: CCCCC(NC(=O)C(NC(=O)C(NC(=O)OCC1=CC=CC=C1)C(C)C)C(C)C)C(=O)C=[N+]=[N-]
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results