accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONHNFFF

CAS RN 155377-19-8

IUPAC Name: ethyl 5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Synonyms: ethyl 3-(trifluoromethyl)-2H-pyrazole-4-carboxylate
Molecular Weight (g/mol): 208.14
Molecular Formula: C7H7F3N2O2
InChi Key: VYXIHSAEOXPAEY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(NN=C1)C(F)(F)F
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results