accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15546-42-6

BrBrPP

CAS RN 15546-42-6

IUPAC Name: triphenyl[4-(triphenylphosphaniumyl)butyl]phosphanium dibromide
Synonyms: triphenyl[4-(triphenylphosphaniumyl)butyl]phosphanium dibromide
Molecular Weight (g/mol): 740.5
Molecular Formula: C40H38Br2P2
InChi Key: WTFXVDWCEPLDDY-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C(CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results