accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15572-56-2

ClH3CH3CNH3

CAS RN 15572-56-2

IUPAC Name: propan-2-aminium chloride
Synonyms: isopropylaminium chloride
Molecular Weight (g/mol): 95.57
Molecular Formula: C3H10ClN
InChi Key: ISYORFGKSZLPNW-UHFFFAOYSA-N
SMILES: [Cl-].CC(C)[NH3+]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results