CAS RN 15574-96-6
IUPAC Name:
1-methyl-4-{6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-2-ylidene}piperidine
Synonyms:
litec polomigran pizotyline sandomigran pizotifenum 苯噻啶 pizotifen sandomygran pizotifen (product) pizotifeno
Molecular Weight (g/mol):
295.44
Molecular Formula:
C19H21NS
InChi Key:
FIADGNVRKBPQEU-UHFFFAOYSA-N
SMILES:
CN1CCC(CC1)=C1C2=C(CCC3=CC=CC=C13)SC=C2
1
–
1
of
1
results
1
Pizotyline, TRC
CAS: 15574-96-6 Molecular Formula: C19 H21 N S Molecular Weight (g/mol): 295.44 Synonym: Piperidine, 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-,4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine,4H-Benzo[4,5]cyclohepta[1,2-b]thiophene, piperidine deriv.,4-(1-Methyl-4-piperidylidene)-9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophene,BC 105,Litec,Pizotifen,Pizotyline,Polomigran,Sandomigran IUPAC Name: 4-(4,5-dihydrobenzo[1,2]cyclohepta[2,4-b]thiophen-10-ylidene)-1-methylpiperidine SMILES: CN1CCC(=C2c3ccccc3CCc4sccc24)CC1