accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Fe(R)PPCH3H3COH

CAS RN 155806-35-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 594.54
Molecular Formula: C36H44FeP2
InChi Key: HGTBZFMPHBAUCQ-UHFFFAOYNA-N
SMILES: [Fe].[CH]1[CH][CH][CH][CH]1.CC([C]1[CH][CH][CH][C]1P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1CCCCC1)C1CCCCC1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)