accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOFFFFFFF

CAS RN 1559-07-5

IUPAC Name: butyl 2,2,3,3,4,4,4-heptafluorobutanoate
Synonyms: butyl 2,2,3,3,4,4,4-heptafluorobutanoate
Molecular Weight (g/mol): 270.15
Molecular Formula: C8H9F7O2
InChi Key: YDXXJZKOOOJVEE-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results