accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3COO

CAS RN 156-54-7

IUPAC Name: sodium butanoate
Synonyms: sodium butyrate butanoic acid, sodium salt butyric acid sodium salt butyric acid, sodium salt butanoic acid, sodium salt (1:1)
Molecular Weight (g/mol): 110.09
Molecular Formula: C4H7NaO2
InChi Key: MFBOGIVSZKQAPD-UHFFFAOYSA-M
SMILES: [Na+].CCCC([O-])=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)