accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HO

CAS RN 156-87-6

IUPAC Name: 3-aminopropan-1-ol
Synonyms: propanolamine
Molecular Weight (g/mol): 75.11
Molecular Formula: C3H9NO
InChi Key: WUGQZFFCHPXWKQ-UHFFFAOYSA-N
SMILES: NCCCO
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (6)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)