accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1560-11-8

HOO

CAS RN 1560-11-8

IUPAC Name: cyclopent-1-ene-1-carboxylic acid
Synonyms: cyclopent-1-ene-1-carboxylic acid
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: PYRZPBDTPRQYKG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)