CAS RN 1562-34-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenyl Ethenesulfonate, TRC
CAS: 1562-34-1 Molecular Formula: C8H8O3S Molecular Weight (g/mol): 184.21 Synonym: Phenyl Ethylenesulfonate,Phenyl Vinylsulfonate IUPAC Name: phenyl ethenesulfonate SMILES: C=CS(=O)(=O)Oc1ccccc1