accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1564-64-3

IUPAC Name: 9-bromoanthracene
Synonyms: 9-bromoanthracene
Molecular Weight (g/mol): 257.13
Molecular Formula: C14H9Br
InChi Key: ZIRVQSRSPDUEOJ-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)