CAS RN 1565-74-8
CAS RN 1565-74-8
IUPAC Name:
1-phenylpropan-1-ol
Synonyms:
1-phenyl-1-propanol phenychol phenycholon livonal ejibil fepar felitrope felicur phenylchol �-ethylbenzyl alcohol
Molecular Weight (g/mol):
136.19
Molecular Formula:
C9H12O
InChi Key:
DYUQAZSOFZSPHD-UHFFFAOYNA-N
SMILES:
CCC(O)C1=CC=CC=C1
1
–
1
of
1
results
1
(R)-(+)-1-Phenyl-1-propanol, 99%
CAS: 1565-74-8 Molecular Formula: C9H12O Molecular Weight (g/mol): 136.194 MDL Number: MFCD00064279 InChI Key: DYUQAZSOFZSPHD-SECBINFHSA-N Synonym: r-+-1-phenyl-1-propanol,r-1-phenylpropan-1-ol,1r-1-phenylpropan-1-ol,r-1-phenyl-1-propanol,r-1-phenyl-propanol,r-+-alpha-ethylbenzyl alcohol,r-+-1-phenylpropanol,r-1-phenylpropanol,1-phenyl-1-propanol #,r-1-phenyl-1propanol PubChem CID: 640199 IUPAC Name: (1R)-1-phenylpropan-1-ol SMILES: CCC(C1=CC=CC=C1)O