accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15656-28-7

IUPAC Name: bis(pyridin-1-ium-1-yl)iodanuide
Synonyms: bis(pyridin-1-ium-1-yl)iodanuide
Molecular Weight (g/mol): 285.11
Molecular Formula: C10H10IN2
InChi Key: BQYJZRLZAKJMMI-UHFFFAOYSA-N
SMILES: [I-]([N+]1=CC=CC=C1)[N+]1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results