accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NClNClPt

CAS RN 15663-27-1

IUPAC Name: cisplatin
Synonyms: cisplatin
Molecular Weight (g/mol): 302.06
Molecular Formula: Cl2H8N2Pt
InChi Key: OJMIKGOOWBVMEV-UHFFFAOYSA-N
SMILES: N.N.Cl.Cl.[Pt]
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)