accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOHOOH

CAS RN 156682-54-1

IUPAC Name: [3-(benzyloxy)phenyl]boronic acid
Synonyms: 3-(benzyloxy)phenylboronic acid
Molecular Weight (g/mol): 228.05
Molecular Formula: C13H13BO3
InChi Key: WIJNYNBSPQMJGO-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(OCC2=CC=CC=C2)=CC=C1
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)