accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 15673-00-4

IUPAC Name: 3,3-dimethylbutan-1-amine
Synonyms: 3,3-dimethylbutan-1-amine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: GPWHFPWZAPOYNO-UHFFFAOYSA-N
SMILES: CC(C)(C)CCN
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results