CAS RN 15686-71-2
IUPAC Name:
hydrogen (6R,7R)-7-[(2R)-2-amino-2-phenylacetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrate chloride
Synonyms:
cefalexin
Molecular Weight (g/mol):
401.86
Molecular Formula:
C16H20ClN3O5S
InChi Key:
YHJDZIQOCSDIQU-OEDJVVDHSA-N
SMILES:
[H+].O.[Cl-].CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=CC=C1)C2=O)C(O)=O
1
–
1
of
1
results
1
Cephalexin hydrate, 97+%
CAS: 15686-71-2 Molecular Formula: C16H20ClN3O5S Molecular Weight (g/mol): 401.862 MDL Number: MFCD00167148 InChI Key: YHJDZIQOCSDIQU-OEDJVVDHSA-N PubChem CID: 57515946 IUPAC Name: (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydron;chloride;hydrate SMILES: [H+].CC1=C(N2C(C(C2=O)NC(=O)C(C3=CC=CC=C3)N)SC1)C(=O)O.O.[Cl-]