accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1569-60-4

CH3HOH3CH3C

CAS RN 1569-60-4

IUPAC Name: 6-methylhept-5-en-2-ol
Synonyms: 6-methyl-5-hepten-2-ol sulcatol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: OHEFFKYYKJVVOX-UHFFFAOYNA-N
SMILES: CC(O)CCC=C(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)