accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFOCl

CAS RN 157033-24-4

IUPAC Name: 2-(3,5-difluorophenyl)acetyl chloride
Synonyms: 2-(3,5-difluorophenyl)acetyl chloride
Molecular Weight (g/mol): 190.57
Molecular Formula: C8H5ClF2O
InChi Key: WMVPARKLYWQLNN-UHFFFAOYSA-N
SMILES: FC1=CC(CC(Cl)=O)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results