accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3

CAS RN 1571-86-4

IUPAC Name: 1,4-dibutylbenzene
Synonyms: 1,4-dibutylbenzene
Molecular Weight (g/mol): 190.33
Molecular Formula: C14H22
InChi Key: VGQOZYOOFXEGDA-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(CCCC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results