accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 157491-15-1

IUPAC Name: N-(2,6-dichlorophenyl)-3-methylbenzamide
Synonyms: N-(2,6-dichlorophenyl)-3-methylbenzamide
Molecular Weight (g/mol): 280.15
Molecular Formula: C14H11Cl2NO
InChi Key: PISANCSDQOSBCO-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)NC1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results