accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOOOOOOOOOCeCe

CAS RN 15750-47-7

IUPAC Name: tris(oxalic acid) dicerium hydrate
Synonyms: tris(oxalic acid) dicerium hydrate
Molecular Weight (g/mol): 568.35
Molecular Formula: C6H8Ce2O13
InChi Key: OGGMXARXHWPYSA-UHFFFAOYSA-N
SMILES: O.[Ce].[Ce].OC(=O)C(O)=O.OC(=O)C(O)=O.OC(=O)C(O)=O
Percent Purity 
  • (2)
  • (4)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)