accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOOHNNH2

CAS RN 1576-35-8

IUPAC Name: 4-methylbenzene-1-sulfonohydrazide
Synonyms: 4-toluenesulfonyl hydrazide
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: ICGLPKIVTVWCFT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NN
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results