accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOOOH

CAS RN 1576-43-8

IUPAC Name: 4-hydroxybenzene-1-sulfonamide
Synonyms: P-hydroxybenzenesulfonamide
Molecular Weight (g/mol): 173.19
Molecular Formula: C6H7NO3S
InChi Key: DIRCLGLKRZLKHG-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(O)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results