accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1576-47-2

IUPAC Name: naphthalene-2-sulfonamide
Synonyms: 2naphthylsulfonamide
Molecular Weight (g/mol): 207.25
Molecular Formula: C10H9NO2S
InChi Key: SWBLLSQMOMPTMC-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results