accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH(S)H3COHOOOH3CCH3H3C

CAS RN 15761-38-3

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 188.2
Molecular Formula: C8H14NO4
InChi Key: QVHJQCGUWFKTSE-YFKPBYRVSA-M
SMILES: C[C@H](NC(=O)OC(C)(C)C)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)