accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 157664-47-6

IUPAC Name: 1-(2-bromothiophen-3-yl)methanamine
Synonyms: 1-(2-bromothiophen-3-yl)methanamine
Molecular Weight (g/mol): 192.07
Molecular Formula: C5H6BrNS
InChi Key: SLKZUJBSIKGHFU-UHFFFAOYSA-N
SMILES: NCC1=C(Br)SC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results