accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOCH2

CAS RN 1577-22-6

IUPAC Name: hex-5-enoic acid
Synonyms: 5-hexenoic acid delta-hexenoic acid
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: XUDOZULIAWNMIU-UHFFFAOYSA-N
SMILES: OC(=O)CCCC=C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)