accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONHN

CAS RN 15788-16-6

IUPAC Name: 1H-1,3-benzodiazole-6-carboxylate
Synonyms: 3H-1,3-benzodiazole-5-carboxylate
Molecular Weight (g/mol): 161.14
Molecular Formula: C8H5N2O2
InChi Key: COYPLDIXZODDDL-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2N=CNC2=C1
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results