accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH3C(E)

CAS RN 15790-88-2

IUPAC Name: methyl (2E)-pent-2-enoate
Synonyms: methyl (2E)-pent-2-enoate
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: MBAHGFJTIVZLFB-SNAWJCMRSA-N
SMILES: CC\C=C\C(=O)OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results