accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFBrF

CAS RN 157911-56-3

IUPAC Name: 1-(bromomethyl)-2,4,5-trifluorobenzene
Synonyms: 1-(bromomethyl)-2,4,5-trifluorobenzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: GAUXEYCSWSMMFZ-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(CBr)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results